missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™

Catalog No. p-7126797
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D157125ML 25 mL
D1571100ML 100 mL
D1571500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D157125ML Supplier TCI America Supplier No. D157125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4246-51-9
Molecular Formula C10H26N2O3
Molecular Weight (g/mol) 222.33
MDL Number MFCD00059850
InChI Key JCEZOHLWDIONSP-UHFFFAOYSA-P
Synonym 4,7,10-trioxa-1,13-tridecanediamine, 3,3'-oxybis ethane-2,1-diyl bis oxy bis propan-1-amine, diethylene glycol bis 3-aminopropyl ether, unii-95w232h4rt, 3,3'-oxybis ethyleneoxy bis propylamine, di 3-aminopropyl ether of diethylene glycol, diethylene glycol, di 3-aminopropyl ether, 1-propanamine, 3,3'-oxybis 2,1-ethanediyloxy bis, 3,3'-oxybis 2,1-ethanediyloxy bis-1-propanamine
PubChem CID 20239
IUPAC Name 3-{2-[2-(3-azaniumylpropoxy)ethoxy]ethoxy}propan-1-aminium
SMILES [NH3+]CCCOCCOCCOCCC[NH3+]

Specifications

Color Yellow
Boiling Point 151°C
Quantity 25 mL
UN Number 2735
Formula Weight 220.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Bis(3-aminopropyl) Ether
EINECSNumber (7)-1234
RTECSNumber ID6475000
TSCA Yes