CAS RN 4246-51-9
IUPAC Name:
3-{2-[2-(3-azaniumylpropoxy)ethoxy]ethoxy}propan-1-aminium
Synonyms:
3-{2-[2-(3-ammoniopropoxy)ethoxy]ethoxy}propan-1-aminium
Molecular Weight (g/mol):
222.33
Molecular Formula:
C10H26N2O3
InChi Key:
JCEZOHLWDIONSP-UHFFFAOYSA-P
SMILES:
[NH3+]CCCOCCOCCOCCC[NH3+]
1
–
1
of
1
results
1
Diethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™
CAS: 4246-51-9 Molecular Formula: C10H26N2O3 Molecular Weight (g/mol): 222.33 MDL Number: MFCD00059850 InChI Key: JCEZOHLWDIONSP-UHFFFAOYSA-P Synonym: 4,7,10-trioxa-1,13-tridecanediamine,3,3'-oxybis ethane-2,1-diyl bis oxy bis propan-1-amine,diethylene glycol bis 3-aminopropyl ether,unii-95w232h4rt,3,3'-oxybis ethyleneoxy bis propylamine,di 3-aminopropyl ether of diethylene glycol,diethylene glycol, di 3-aminopropyl ether,1-propanamine, 3,3'-oxybis 2,1-ethanediyloxy bis,3,3'-oxybis 2,1-ethanediyloxy bis-1-propanamine PubChem CID: 20239 IUPAC Name: 3-{2-[2-(3-azaniumylpropoxy)ethoxy]ethoxy}propan-1-aminium SMILES: [NH3+]CCCOCCOCCOCCC[NH3+]