missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Bis(2-propynyl) Ether 98.0+%, TCI America™
$303.84 - $1051.33
Chemical Identifiers
| CAS | 126422-57-9 |
|---|---|
| Molecular Formula | C10H14O3 |
| Molecular Weight (g/mol) | 182.219 |
| InChI Key | ZQNBDEOGKNZIQM-UHFFFAOYSA-N |
| Synonym | 3-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]-1-propyne, 4,7,10-Trioxa-1,12-tridecadiyne |
| PubChem CID | 11008566 |
| IUPAC Name | 3-[2-(2-prop-2-ynoxyethoxy)ethoxy]prop-1-yne |
| SMILES | C#CCOCCOCCOCC#C |
Chemical Identifiers
| 126422-57-9 | |
| 182.219 | |
| 3-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]-1-propyne, 4,7,10-Trioxa-1,12-tridecadiyne | |
| 3-[2-(2-prop-2-ynoxyethoxy)ethoxy]prop-1-yne |
| C10H14O3 | |
| ZQNBDEOGKNZIQM-UHFFFAOYSA-N | |
| 11008566 | |
| C#CCOCCOCCOCC#C |
Specifications
| 126422-57-9 | |
| 103°C | |
| 5 g | |
| ZQNBDEOGKNZIQM-UHFFFAOYSA-N | |
| 3-[2-(2-prop-2-ynoxyethoxy)ethoxy]prop-1-yne | |
| 11008566 | |
| ≥98.0% (GC) | |
| Diethylene Glycol Bis(2-propynyl) Ether |
| Yellow | |
| C10H14O3 | |
| 3-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]-1-propyne, 4,7,10-Trioxa-1,12-tridecadiyne | |
| C#CCOCCOCCOCC#C | |
| 182.219 | |
| 182.22 | |
| Liquid |
Safety and Handling
TSCA : No