accessibility menu, dialog, popup

UserName

CAS RN 126422-57-9

HCOOOCH

CAS RN 126422-57-9

IUPAC Name: 3-{2-[2-(prop-2-yn-1-yloxy)ethoxy]ethoxy}prop-1-yne
Synonyms: 3-{2-[2-(prop-2-yn-1-yloxy)ethoxy]ethoxy}prop-1-yne
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: ZQNBDEOGKNZIQM-UHFFFAOYSA-N
SMILES: C#CCOCCOCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results