missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Bis(2-chloroethyl) Ether 97.0+%, TCI America™

Catalog No. D1784500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1784500G 500 g
D178425G 25 g
D1784100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1784500G Supplier TCI America Supplier No. D1784500G

Chemical Identifiers

CAS 638-56-2
Molecular Formula C8H16Cl2O3
Molecular Weight (g/mol) 231.113
MDL Number MFCD00054473
InChI Key ZCFRYTWBXNQVOW-UHFFFAOYSA-N
PubChem CID 61187
IUPAC Name 1-(2-chloroethoxy)-2-[2-(2-chloroethoxy)ethoxy]ethane
SMILES C(COCCCl)OCCOCCCl

Specifications

Color Yellow
Quantity 500 g
UN Number 2810
Formula Weight 231.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Bis(2-chloroethyl) Ether
RTECSNumber KN0525000
TSCA No