accessibility menu, dialog, popup

UserName

ClOOOCl

CAS RN 638-56-2

IUPAC Name: 1-chloro-2-{2-[2-(2-chloroethoxy)ethoxy]ethoxy}ethane
Synonyms: tetraglycol dichloride
Molecular Weight (g/mol): 231.11
Molecular Formula: C8H16Cl2O3
InChi Key: ZCFRYTWBXNQVOW-UHFFFAOYSA-N
SMILES: ClCCOCCOCCOCCCl
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results