Learn More
Diethylene glycol, 99%
CAS: 111-46-6 | C4H10O3 | 106.12 g/mol
Supplier: Thermo Scientific Chemicals A147280E
Description
Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.
Solubility
Miscible with water, alcohol, ether, acetone, and ethylene glycol.
Notes
Hygroscopic. Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and zinc.
Specifications
| Diethylene glycol | |
| -10°C | |
| 245°C to 246°C | |
| 99% | |
| C4H10O3 | |
| (HOCH2CH2)2O | |
| 2500 g | |
| Hygroscopic | |
| diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
| MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
| 2-(2-hydroxyethoxy)ethanol | |
| 8117 | |
| 106.12 | |
| Liquid |
| 111-46-6 | |
| 1.118 | |
| 143°C (289°F) | |
| Nearly Odorless | |
| 1.447 | |
| MFCD00002882 | |
| 969209 | |
| 14,3119 | |
| Miscible with water,alcohol,ether,acetone,and ethylene glycol. | |
| OCCOCCO | |
| 106.12 | |
| CHEBI:46807 | |
| 99% |
Chemical Identifiers
| 111-46-6 | |
| 106.12 | |
| MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
| 8117 | |
| OCCOCCO |
| C4H10O3 | |
| MFCD00002882 | |
| diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
| CHEBI:46807 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P260-P264b-P270-P301+P312-P314-P330-P501c
H302-H373
EINECSNumber : 203-872-2
RTECSNumber : ID5950000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only