missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012469
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012469 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012469 Supplier Merck Emd Millipore Supplier No. 031285MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diethylene glycol is a colourless liquid which is mostly used in manufacturing other chemicals. It is denser than water. It can be formed as a side reaction during ester interchange of dimethyl terephthalate with ethylene glycol, it can also be formed when terephthalic acid undergoes direct esterification with ethylene glycol. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Description Non-ionic
CAS 111-46-6
Ignition Point 442°F
Vapor Pressure 0.01 mmHg (20°C)
Vapor Density 2.14 (vs. air)
Density 1.118 g/mL (at 25°C)
Flash Point 143°C
Physical Form Viscous Liquid
Linear Formula (HOCH2 CH2)2 O
MDL Number MFCD00002882
Melting Point -10°C
Boiling Point 245°C
Synonym 2,2′-Oxydiethanol; 2-Hydroxyethyl ether; Bis(2-hydroxyethyl) ether; Diglycol
Type Analytical Standard
Quantity 5 mL
Formula Weight 106.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less