missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D102720LT

 View more versions of this product

Catalog No. 18610193


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

111-46-6
Poly Drum
0.97
1
20 L
Reagent

Chemical Identifiers

111-46-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.