missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol, 97%, Spectrum™ Chemical

Catalog No. 18610192
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610192 1 L Amber Glass Bottle
18610195 4 L Amber Glass Bottle
18610193 20 L Amber Glass Bottle
18610194 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610192 Supplier Spectrum Chemical Mfg Cor Supplier No. D10271LT
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 111-46-6

Specifications

CAS Max % 1
Boiling Point 245°C to 246°C (473°F to 475°F)
Packaging Amber Glass Bottle
Quantity 1 L
Molecular Weight (g/mol) 106.12
Percent Purity 0.97
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Diethylene Glycol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.