Learn More
Diethylbenzene, mixture of isomers
CAS: 25340-17-4 | C10H14
$41.66 - $225.96
Chemical Identifiers
| CAS | 25340-17-4 |
|---|---|
| Molecular Formula | C10H14 |
| MDL Number | MFCD00792900 |
| Synonym | Dowex J |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA19548AE
|
Thermo Scientific Chemicals
A19548AE |
100 mL |
Each for $41.66
|
|
|||||
|
AAA19548AP
|
Thermo Scientific Chemicals
A19548AP |
500 mL |
Each for $71.14
|
|
|||||
|
AAA195480F
|
Thermo Scientific Chemicals
A195480F |
2500 mL |
Each for $225.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25340-17-4 | |
| C10H14 | |
| MFCD00792900 | |
| Dowex J |
Specifications
| 25340-17-4 | |
| 182°C to 184°C | |
| C10H14 | |
| MFCD00792900 | |
| UN2049 | |
| Dowex J | |
| Diethylbenzene, mixture of isomers |
| 0.87 | |
| 55°C (131°F) | |
| 1.495 | |
| 100 mL | |
| 1903396 | |
| 134.22 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H226-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber : 246-874-9
RTECSNumber : CZ5600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only