CAS RN 25340-17-4
CAS RN 25340-17-4
IUPAC Name:
1,2-diethylbenzene
Synonyms:
diethylbenzenes O-diethylbenzene
Molecular Weight (g/mol):
134.22
Molecular Formula:
C10H14
InChi Key:
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILES:
CCC1=CC=CC=C1CC
1
–
5
of
5
results
1
Diethylbenzene, mixture of isomers
CAS: 25340-17-4 Molecular Formula: C10H14 MDL Number: MFCD00792900 Synonym: Dowex J
2
Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%
CAS: 25340-17-4 Molecular Formula: C10H14 Molecular Weight (g/mol): 134.22 MDL Number: MFCD00792900
4
Diethylbenzene (mixture) 70.0+%, TCI America™
CAS: 25340-17-4 Molecular Formula: C10H14 MDL Number: MFCD00792900
5
Diethylbenzene (mixture) 98.0+%, TCI America™
CAS: 25340-17-4 Molecular Formula: C10H14 MDL Number: MFCD00792900