missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylammonium Diethyldithiocarbamate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 222.41 |
| Formula Weight | 222.41 |
| Percent Purity | ≥97.0% (T) |
| Grade | Laboratory |
| Chemical Name or Material | Diethylammonium Diethyldithiocarbamate |
Safety and Handling
| EINECSNumber | (2)-2894 |
| RTECSNumber | EZ5425000 |
| TSCA | Yes |