missing translation for 'onlineSavingsMsg'
Learn More

Diethylammonium diethyldithiocarbamate, 98%

CAS: 1518-58-7 | C9H22N2S2

Supplier:  Thermo Scientific Chemicals A1045814

 View more versions of this product

Catalog No. AAA1045814


Only null left
Explore available promotions

Description

Description

Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
Specifications

Specifications

Diethylammonium diethyldithiocarbamate
81°C to 84°C
(CH3CH2)2NH2+ (CH3CH2)2NCS2-
25 g
Soluble in Chloroform,Methanol.
98%
1518-58-7
C9H22N2S2
MFCD00013146
Hygroscopic
222.42

Chemical Identifiers

1518-58-7
C9H22N2S2
MFCD00013146
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-175-3

RTECSNumber : EZ5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only