missing translation for 'onlineSavingsMsg'
Learn More

Diethylaminosulfur trifluoride, 95%

Catalog No. p-7081821
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1199206 5 g
AAA1199214 25 g
AAA1199222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1199206 Supplier Thermo Scientific Chemicals Supplier No. A1199206
Only null left

CAS: 38078-09-0 | C4H10F3NS | 161.186 g/mol

Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

Solubility
Miscible with water, ethanol, benzene and acetonitrile.

Notes
Store in cool place. Moisture sensitive. It reacts with water and ethanol. Incompatible with water, acids and bases.

Chemical Identifiers

CAS 38078-09-0
Molecular Formula C4H10F3NS
Molecular Weight (g/mol) 161.186
MDL Number MFCD00000363
InChI Key CSJLBAMHHLJAAS-UHFFFAOYSA-N
Synonym diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
PubChem CID 123472
IUPAC Name N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
SMILES CCN(CC)S(F)(F)F

Specifications

Density 1.23
Boiling Point 30°C to 32°C (3 mmHg)
Flash Point 23°C (73°F)
Refractive Index 1.4115
Linear Formula (CH3CH2)2NSF3
Quantity 5 g
UN Number UN2920
Beilstein 1849066
Sensitivity Moisture sensitive
Solubility Information Miscible with water,ethanol,benzene and acetonitrile.
Formula Weight 161.19
Percent Purity 95%
Chemical Name or Material Diethylaminosulfur trifluoride
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H226
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 253-771-2
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only