Learn More
Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
$147.30 - $147.30
Chemical Identifiers
| CAS | 2040-00-8 |
|---|---|
| Molecular Formula | C4H10AlI |
| Molecular Weight (g/mol) | 212.01 |
| MDL Number | MFCD00001025 |
| InChI Key | PPQUYYAZSOKTQD-UHFFFAOYSA-M |
| Synonym | aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali |
| PubChem CID | 74874 |
| IUPAC Name | diethyl(iodo)alumane |
| SMILES | CC[Al](CC)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC381161000
|
Thermo Scientific Chemicals
381161000 |
100 mL |
Each for $147.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
| 2040-00-8 | |
| 212.01 | |
| PPQUYYAZSOKTQD-UHFFFAOYSA-M | |
| 74874 | |
| CC[Al](CC)I |
| C4H10AlI | |
| MFCD00001025 | |
| aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali | |
| diethyl(iodo)alumane |
Specifications
| 2040-00-8,108-88-3 | |
| AcroSeal™ Glass Bottle | |
| (C2H5)2AlI | |
| 100 mL | |
| Solubility in water: reacts | |
| CC[Al](CC)I | |
| 212.01 | |
| 212.01 | |
| Diethylaluminium iodide, 0.8M Solution in Toluene |
| Colorless | |
| C4H10AlI | |
| MFCD00001025 | |
| aluminum, diethyliodo, diethylaluminum iodide, diethylaluminium iodide, diethyl iodo alumane, et2ali | |
| PPQUYYAZSOKTQD-UHFFFAOYSA-M | |
| diethyl(iodo)alumane | |
| 74874 | |
| Liquid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Reacts violently with water.
GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 218-032-
RUO – Research Use Only