accessibility menu, dialog, popup

UserName

CAS RN 2040-00-8

IUPAC Name: diethyl(iodo)alumane
Synonyms: diethylaluminium iodide
Molecular Weight (g/mol): 212.01
Molecular Formula: C4H10AlI
InChi Key: PPQUYYAZSOKTQD-UHFFFAOYSA-M
SMILES: CC[Al](I)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results