missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminium chloride, 1M solution in hexanes, AcroSeal™

CAS: 96-10-6 | C4H10AlCl | 120.56 g/mol

$163.77 - $163.77

Chemical Identifiers

CAS 96-10-6
Molecular Formula C4H10AlCl
Molecular Weight (g/mol) 120.56
MDL Number MFCD00000459
InChI Key YNLAOSYQHBDIKW-UHFFFAOYSA-M
Synonym diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
PubChem CID 7277
IUPAC Name chloro(diethyl)alumane
SMILES CC[Al](Cl)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199521000
SDS View Documents
Thermo Scientific Chemicals
199521000
100 mL AcroSeal™ Glass bottle
Each for $163.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

96-10-6
120.56
YNLAOSYQHBDIKW-UHFFFAOYSA-M
7277
CC[Al](Cl)CC
C4H10AlCl
MFCD00000459
diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
chloro(diethyl)alumane

Specifications

Colorless
C4H10AlCl
MFCD00000459
0.7110g/mL
Solubility in water: reacts.
CC[Al](Cl)CC
120.56
120.56
0.711
−22°C
Diethylaluminium chloride, 1M Solution in Hexanes
96-10-6,110-54-3
(C2H5)2AICl
0.95 to 1.10M
diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
YNLAOSYQHBDIKW-UHFFFAOYSA-M
chloro(diethyl)alumane
7277
AcroSeal™ Glass bottle
100 mL
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container.

Danger

EINECSNumber : 202-477-2

RTECSNumber : BD0558000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only