missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tetrafluorosuccinate, 97%

Catalog No. AAL1646903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1646903 1 g
AAL1646906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1646903 Supplier Thermo Scientific Chemicals Supplier No. L1646903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 377-71-9 | C8H10F4O4 | 246.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 377-71-9
Molecular Formula C8H10F4O4
Molecular Weight (g/mol) 246.158
MDL Number MFCD00015155
InChI Key BLZSSBPZHBFNBN-UHFFFAOYSA-N
Synonym diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
PubChem CID 67834
IUPAC Name diethyl 2,2,3,3-tetrafluorobutanedioate
SMILES CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F

Specifications

Density 1.273
Boiling Point 78°C (5 mmHg)
Refractive Index 1.368
Quantity 1 g
Beilstein 1798175
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material Diethyl tetrafluorosuccinate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-821-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only