missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfide, 96%

Catalog No. AAL0349309
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0349309 10 g
AAL0349322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0349309 Supplier Thermo Scientific Chemicals Supplier No. L0349309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 352-93-2 | C4H10S | 90.18 g/mol

Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

Solubility
Miscible with alcohol, ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 352-93-2
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00009270
InChI Key LJSQFQKUNVCTIA-UHFFFAOYSA-N
Synonym diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
PubChem CID 9609
ChEBI CHEBI:27710
IUPAC Name ethylsulfanylethane
SMILES CCSCC

Specifications

Melting Point -104°C
Density 0.835
Boiling Point 90°C to 92°C
Flash Point −10°C (14°F)
Odor Garlic-like
Refractive Index 1.442
Quantity 10 g
UN Number UN2375
Beilstein 1696909
Merck Index 14,3854
Solubility Information Miscible with alcohol,ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 90.19
Percent Purity 96%
Chemical Name or Material Diethyl sulfide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIETHYL SULFIDE
EINECSNumber 206-526-9
RTECSNumber LC7200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only