missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Sebacate 98.0+%, TCI America™
$50.33 - $219.95
Chemical Identifiers
| CAS | 110-40-7 |
|---|---|
| Molecular Formula | C14H26O4 |
| Molecular Weight (g/mol) | 258.358 |
| MDL Number | MFCD00009219 |
| InChI Key | ONKUXPIBXRRIDU-UHFFFAOYSA-N |
| Synonym | diethyl sebacate, ethyl sebacate, decanedioic acid, diethyl ester, bisoflex des, diethyl 1,10-decanedioate, sebacic acid, diethyl ester, ethyl sebacate van, sebacic acid diethyl ester, diethyl 1,8-octanedicarboxylate, unii-i41b9fjk6v |
| PubChem CID | 8049 |
| IUPAC Name | diethyl decanedioate |
| SMILES | CCOC(=O)CCCCCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S002425ML
|
TCI America
S002425ML |
25 mL |
Each for $50.33
|
|
|||||
|
S0024500ML
|
TCI America
S0024500ML |
500 mL |
Each for $219.95
|
|
|||||
Chemical Identifiers
| 110-40-7 | |
| 258.358 | |
| ONKUXPIBXRRIDU-UHFFFAOYSA-N | |
| 8049 | |
| CCOC(=O)CCCCCCCCC(=O)OCC |
| C14H26O4 | |
| MFCD00009219 | |
| diethyl sebacate, ethyl sebacate, decanedioic acid, diethyl ester, bisoflex des, diethyl 1,10-decanedioate, sebacic acid, diethyl ester, ethyl sebacate van, sebacic acid diethyl ester, diethyl 1,8-octanedicarboxylate, unii-i41b9fjk6v | |
| diethyl decanedioate |
Specifications
| 110-40-7 | |
| 309°C | |
| MFCD00009219 | |
| diethyl sebacate, ethyl sebacate, decanedioic acid, diethyl ester, bisoflex des, diethyl 1,10-decanedioate, sebacic acid, diethyl ester, ethyl sebacate van, sebacic acid diethyl ester, diethyl 1,8-octanedicarboxylate, unii-i41b9fjk6v | |
| CCOC(=O)CCCCCCCCC(=O)OCC | |
| 258.358 | |
| 258.36 | |
| Liquid |
| Yellow | |
| C14H26O4 | |
| 25 mL | |
| ONKUXPIBXRRIDU-UHFFFAOYSA-N | |
| diethyl decanedioate | |
| 8049 | |
| ≥98.0% (GC) | |
| Diethyl Sebacate |
Safety and Handling
EINECSNumber : (2)-0879
RTECSNumber : VS1180000
TSCA : Yes