missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Sebacate 98.0+%, TCI America™

Catalog No. S0024500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0024500ML 500 mL
S002425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0024500ML Supplier TCI America Supplier No. S0024500ML

Chemical Identifiers

CAS 110-40-7
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00009219
InChI Key ONKUXPIBXRRIDU-UHFFFAOYSA-N
Synonym diethyl sebacate, ethyl sebacate, decanedioic acid, diethyl ester, bisoflex des, diethyl 1,10-decanedioate, sebacic acid, diethyl ester, ethyl sebacate van, sebacic acid diethyl ester, diethyl 1,8-octanedicarboxylate, unii-i41b9fjk6v
PubChem CID 8049
IUPAC Name diethyl decanedioate
SMILES CCOC(=O)CCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 309°C
Quantity 500 mL
Formula Weight 258.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Sebacate
EINECSNumber (2)-0879
RTECSNumber VS1180000
TSCA Yes