missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (S)-(-)-2-Isocyanatoglutarate 97.0+%, TCI America™
$142.46 - $473.59
Chemical Identifiers
| CAS | 145080-95-1 |
|---|---|
| Molecular Formula | C10H15NO5 |
| Molecular Weight (g/mol) | 229.232 |
| MDL Number | MFCD00191663 |
| InChI Key | TZTLQVZEVNEAQD-QMMMGPOBSA-N |
| Synonym | Diethyl (S)-(-)-2-Isocyanatopentanedioate, (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester |
| PubChem CID | 10398920 |
| IUPAC Name | diethyl (2S)-2-isocyanatopentanedioate |
| SMILES | CCOC(=O)CCC(C(=O)OCC)N=C=O |
Chemical Identifiers
| 145080-95-1 | |
| 229.232 | |
| TZTLQVZEVNEAQD-QMMMGPOBSA-N | |
| 10398920 | |
| CCOC(=O)CCC(C(=O)OCC)N=C=O |
| C10H15NO5 | |
| MFCD00191663 | |
| Diethyl (S)-(-)-2-Isocyanatopentanedioate, (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester | |
| diethyl (2S)-2-isocyanatopentanedioate |
Specifications
| 145080-95-1 | |
| 151°C | |
| MFCD00191663 | |
| Diethyl (S)-(-)-2-Isocyanatopentanedioate, (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester | |
| CCOC(=O)CCC(C(=O)OCC)N=C=O | |
| 229.232 | |
| 229.23 | |
| Liquid |
| Yellow | |
| C10H15NO5 | |
| 1 g | |
| TZTLQVZEVNEAQD-QMMMGPOBSA-N | |
| diethyl (2S)-2-isocyanatopentanedioate | |
| 10398920 | |
| ≥97.0% (GC) | |
| Diethyl (S)-(-)-2-Isocyanatoglutarate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator