accessibility menu, dialog, popup

UserName

H3COOOOCH3NCO

CAS RN 145080-95-1

IUPAC Name: 1,5-diethyl 2-isocyanatopentanedioate
Synonyms: 1,5-diethyl 2-isocyanatopentanedioate
Molecular Weight (g/mol): 229.23
Molecular Formula: C10H15NO5
InChi Key: TZTLQVZEVNEAQD-UHFFFAOYNA-N
SMILES: CCOC(=O)CCC(N=C=O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results