missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (S)-(-)-2-Isocyanatoglutarate 97.0+%, TCI America™

Catalog No. I04681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04681G 1 g
I04685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04681G Supplier TCI America Supplier No. I04681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 145080-95-1
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.232
MDL Number MFCD00191663
InChI Key TZTLQVZEVNEAQD-QMMMGPOBSA-N
Synonym Diethyl (S)-(-)-2-Isocyanatopentanedioate, (S)-(-)-2-Isocyanatoglutaric Acid Diethyl Ester, (S)-(-)-2-Isocyanatopentanedioic Acid Diethyl Ester
PubChem CID 10398920
IUPAC Name diethyl (2S)-2-isocyanatopentanedioate
SMILES CCOC(=O)CCC(C(=O)OCC)N=C=O

Specifications

Color Yellow
Boiling Point 151°C
Quantity 1 g
Formula Weight 229.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (S)-(-)-2-Isocyanatoglutarate
TSCA No
Recommended Storage Refrigerator