missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Pimelate 97.0+%, TCI America™
Supplier: TCI America P110025ML
Specifications
| Diethyl Pimelate | |
| Yellow | |
| C11H20O4 | |
| 25 mL | |
| LKKOGZVQGQUVHF-UHFFFAOYSA-N | |
| diethyl heptanedioate | |
| 16300 | |
| ≥97.0% (GC) |
| 2050-20-6 | |
| 149°C | |
| MFCD00009216 | |
| diethyl pimelate, heptanedioic acid, diethyl ester, diethylpimelate, pimelic acid diethyl ester, unii-2rf63n709k, heptanedioic acid diethyl ester, heptanedioic acid, 1,7-diethyl ester, 1,5-pentanedicarboxylic acid diethyl ester, zlchem 330 | |
| CCOC(=O)CCCCCC(=O)OCC | |
| 216.277 | |
| 216.28 | |
| Liquid |
Chemical Identifiers
| 2050-20-6 | |
| 216.277 | |
| LKKOGZVQGQUVHF-UHFFFAOYSA-N | |
| 16300 | |
| CCOC(=O)CCCCCC(=O)OCC |
| C11H20O4 | |
| MFCD00009216 | |
| diethyl pimelate, heptanedioic acid, diethyl ester, diethylpimelate, pimelic acid diethyl ester, unii-2rf63n709k, heptanedioic acid diethyl ester, heptanedioic acid, 1,7-diethyl ester, 1,5-pentanedicarboxylic acid diethyl ester, zlchem 330 | |
| diethyl heptanedioate |
Safety and Handling
RTECSNumber : UV9702000
TSCA : No