accessibility menu, dialog, popup

UserName

H3COOOOCH3

CAS RN 2050-20-6

IUPAC Name: 1,7-diethyl heptanedioate
Synonyms: diethyl pimelate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: LKKOGZVQGQUVHF-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results