missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (Phthalimidomethyl)phosphonate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 63°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 297.25 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Diethyl (Phthalimidomethyl)phosphonate |
Safety and Handling
| TSCA | No |