accessibility menu, dialog, popup

UserName

CAS RN 33512-26-4

IUPAC Name: diethyl [(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phosphonate
Synonyms: diethyl (1,3-dioxoisoindol-2-yl)methylphosphonate
Molecular Weight (g/mol): 297.25
Molecular Formula: C13H16NO5P
InChi Key: IUZMHUAHKBHJFY-UHFFFAOYSA-N
SMILES: CCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)