missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Phthalimidomethyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7134998
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11935G 5 g
P119325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11935G Supplier TCI America Supplier No. P11935G

Chemical Identifiers

CAS 33512-26-4
Molecular Formula C13H16NO5P
Molecular Weight (g/mol) 297.247
MDL Number MFCD00134313
InChI Key IUZMHUAHKBHJFY-UHFFFAOYSA-N
Synonym diethyl phthalimidomethyl phosphonate, diethyl 1,3-dioxoisoindolin-2-yl methyl phosphonate, phthalimidomethyl phosphonic acid diethyl ester, diethyl 1,3-dioxoisoindol-2-yl methylphosphonate, 2-diethoxycarbonyl methyl benzo c azoline-1,3-dione, acmc-1cp9b, diethyl phthalimidomethylphosphonate, n-diethoxyphosphoryl methyl phthalimide
PubChem CID 1924456
IUPAC Name 2-(diethoxyphosphorylmethyl)isoindole-1,3-dione
SMILES CCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC

Specifications

Melting Point 63°C
Color White
Quantity 5 g
Formula Weight 297.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl (Phthalimidomethyl)phosphonate