missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (phthalimidomethyl)phosphonate, 97%

Catalog No. AAB2109606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2109606 5 g
AAB2109614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2109606 Supplier Thermo Scientific Chemicals Supplier No. B2109606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33512-26-4 | C13H16NO5P | 297.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33512-26-4
Molecular Formula C13H16NO5P
Molecular Weight (g/mol) 297.247
MDL Number MFCD00134313
InChI Key IUZMHUAHKBHJFY-UHFFFAOYSA-N
Synonym diethyl phthalimidomethyl phosphonate, diethyl 1,3-dioxoisoindolin-2-yl methyl phosphonate, phthalimidomethyl phosphonic acid diethyl ester, diethyl 1,3-dioxoisoindol-2-yl methylphosphonate, 2-diethoxycarbonyl methyl benzo c azoline-1,3-dione, acmc-1cp9b, diethyl phthalimidomethylphosphonate, n-diethoxyphosphoryl methyl phthalimide
PubChem CID 1924456
IUPAC Name 2-(diethoxyphosphorylmethyl)isoindole-1,3-dione
SMILES CCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Formula Weight 297.25
Percent Purity 97%
Chemical Name or Material Diethyl (phthalimidomethyl)phosphonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only