missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate 99.0+%, TCI America™

Supplier:  TCI America P0296500ML

 View more versions of this product

Catalog No. P0296500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Diethyl Phthalate
-41°C
296°C
MFCD00009111
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
CCOC(=O)C1=CC=CC=C1C(=O)OCC
222.24
CHEBI:34698
≥99.0% (GC)
84-66-2
Colorless
C12H14O4
500 mL
FLKPEMZONWLCSK-UHFFFAOYSA-N
diethyl benzene-1,2-dicarboxylate
6781
222.24
Liquid

Chemical Identifiers

84-66-2
222.24
FLKPEMZONWLCSK-UHFFFAOYSA-N
6781
diethyl benzene-1,2-dicarboxylate
C12H14O4
MFCD00009111
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
CHEBI:34698
CCOC(=O)C1=CC=CC=C1C(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1301

RTECSNumber : TI1050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions