missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate 99.0+%, TCI America™

Catalog No. P029625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P029625ML 25 mL
P0296500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P029625ML Supplier TCI America Supplier No. P029625ML

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Melting Point -41°C
Color Colorless
Boiling Point 296°C
Quantity 25 mL
Formula Weight 222.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Phthalate
EINECSNumber (3)-1301
RTECSNumber TI1050000
TSCA Yes