missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111018271
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018271 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018271 Supplier Merck Emd Millipore Supplier No. 3418525MG
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 93952-12-6
Physical Form Neat
Linear Formula C6D4(CO2C2H5)2
MDL Number MFCD00144117
Melting Point -3°C
Boiling Point 298°C to 299°C
Type Analytical Standard
Quantity 25 mg
Formula Weight 226.26
Grade Analytical Standard