missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Phenyl Orthoformate 97.0+%, TCI America™
Supplier: TCI America O018725G
Specifications
| Diethyl Phenyl Orthoformate | |
| -26°C | |
| 104°C | |
| MFCD00009228 | |
| diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester | |
| CCOC(OCC)OC1=CC=CC=C1 | |
| 196.246 | |
| 196.25 | |
| Liquid |
| 14444-77-0 | |
| Colorless | |
| C11H16O3 | |
| 25 g | |
| QTURWMMVIIBRRP-UHFFFAOYSA-N | |
| diethoxymethoxybenzene | |
| 84440 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 14444-77-0 | |
| 196.246 | |
| QTURWMMVIIBRRP-UHFFFAOYSA-N | |
| 84440 | |
| CCOC(OCC)OC1=CC=CC=C1 |
| C11H16O3 | |
| MFCD00009228 | |
| diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester | |
| diethoxymethoxybenzene |
Safety and Handling
TSCA : No