missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phenyl Orthoformate 97.0+%, TCI America™

Catalog No. p-7134231
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01875G 5 g
O018725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01875G Supplier TCI America Supplier No. O01875G
Only null left

Chemical Identifiers

CAS 14444-77-0
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00009228
InChI Key QTURWMMVIIBRRP-UHFFFAOYSA-N
Synonym diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester
PubChem CID 84440
IUPAC Name diethoxymethoxybenzene
SMILES CCOC(OCC)OC1=CC=CC=C1

Specifications

Melting Point -26°C
Color Colorless
Boiling Point 104°C
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Phenyl Orthoformate