missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals

Catalog No. AAH3769706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3769706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3769706 Supplier Thermo Scientific Chemicals Supplier No. H3769706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4844-38-6 | Molecular Formula: C16H35O3P | Molecular Weight: 306.427 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4844-38-6
Molecular Formula C16H35O3P
Molecular Weight (g/mol) 306.427
MDL Number MFCD00015595
InChI Key LRIWHHSVZCKNJL-UHFFFAOYSA-N
Synonym diethyl dodecylphosphonate, phosphonic acid, dodecyl-, diethyl ester, diethyl 1-dodecyl phosphonate, phosphonic acid, p-dodecyl-, diethyl ester, dodecylphosphonic acid diethyl ester, phosphonic acid,p-dodecyl-, diethyl ester
PubChem CID 78558
IUPAC Name 1-diethoxyphosphoryldodecane
SMILES CCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 168°C (1.5 mmHg)
Quantity 5 g
Formula Weight 306.42
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Diethyl n-dodecylphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-429-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only