CAS RN 4844-38-6
IUPAC Name:
diethyl dodecylphosphonate
Synonyms:
diethyl dodecylphosphonate
Molecular Weight (g/mol):
306.43
Molecular Formula:
C16H35O3P
InChi Key:
LRIWHHSVZCKNJL-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCP(=O)(OCC)OCC
1
–
1
of
1
results
1
Promotions Available
Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals
Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals
CAS: 4844-38-6 Molecular Formula: C16H35O3P Molecular Weight (g/mol): 306.427 MDL Number: MFCD00015595 InChI Key: LRIWHHSVZCKNJL-UHFFFAOYSA-N Synonym: diethyl dodecylphosphonate,phosphonic acid, dodecyl-, diethyl ester,diethyl 1-dodecyl phosphonate,phosphonic acid, p-dodecyl-, diethyl ester,dodecylphosphonic acid diethyl ester,phosphonic acid,p-dodecyl-, diethyl ester PubChem CID: 78558 IUPAC Name: 1-diethoxyphosphoryldodecane SMILES: CCCCCCCCCCCCP(=O)(OCC)OCC