missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Catalog No. AAA1477218
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1477218 50 g
AAA1477209 10 g
AAA1477214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1477218 Supplier Thermo Scientific Chemicals Supplier No. A1477218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 683-08-9
Molecular Formula C5H13O3P
Molecular Weight (g/mol) 152.13
MDL Number MFCD00009813
InChI Key NYYLZXREFNYPKB-UHFFFAOYSA-N
Synonym diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h
PubChem CID 12685
IUPAC Name 1-[ethoxy(methyl)phosphoryl]oxyethane
SMILES CCOP(C)(=O)OCC

Specifications

Density 1.04
Boiling Point 194°C
Flash Point 75°C (167°F)
Refractive Index 1.414
Quantity 50 g
Beilstein 1753416
Solubility Information Miscible with water.
Formula Weight 152.13
Percent Purity 97%
Chemical Name or Material Diethyl methylphosphonate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-667-4
RTECSNumber SZ9085000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only