accessibility menu, dialog, popup

UserName

CAS RN 683-08-9

IUPAC Name: diethyl methylphosphonate
Synonyms: diethyl methylphosphonate
Molecular Weight (g/mol): 152.13
Molecular Formula: C5H13O3P
InChi Key: NYYLZXREFNYPKB-UHFFFAOYSA-N
SMILES: CCOP(C)(=O)OCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)