missing translation for 'onlineSavingsMsg'
Learn More

Diethyl(methyl)propylammonium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Catalog No. D52371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D52371G Supplier TCI America Supplier No. D52371G

Chemical Identifiers

CAS 1235234-32-8
Molecular Formula C8H20F2N2O4S2
Molecular Weight (g/mol) 310.375
InChI Key JVCJTUSCMOJHBG-UHFFFAOYSA-N
PubChem CID 121234257
IUPAC Name bis(fluorosulfonyl)azanide;diethyl-methyl-propylazanium
SMILES CCC[N+](C)(CC)CC.[N-](S(=O)(=O)F)S(=O)(=O)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 310.37
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material Diethyl(methyl)propylammonium Bis(fluorosulfonyl)imide