accessibility menu, dialog, popup

UserName

CH3NCH3H3CH3CNSOOFSOOF

CAS RN 1235234-32-8

IUPAC Name: bis(fluorosulfonyl)azanide; diethyl(methyl)propylazanium
Synonyms: bis(fluorosulfonyl)azanide; diethyl(methyl)propylazanium
Molecular Weight (g/mol): 310.37
Molecular Formula: C8H20F2N2O4S2
InChi Key: JVCJTUSCMOJHBG-UHFFFAOYSA-N
SMILES: CCC[N+](C)(CC)CC.FS(=O)(=O)[N-]S(F)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results