missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (Methoxymethyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7127967
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38735G 5 g
D387325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38735G Supplier TCI America Supplier No. D38735G
Only null left

Chemical Identifiers

CAS 32806-04-5
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00137944
InChI Key IWEOGHGFCXMKFM-UHFFFAOYSA-N
Synonym (Methoxymethyl)phosphonic Acid Diethyl Ester
PubChem CID 11658394
IUPAC Name diethyl (methoxymethyl)phosphonate
SMILES CCOP(=O)(COC)OCC

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
Formula Weight 182.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (Methoxymethyl)phosphonate