accessibility menu, dialog, popup

UserName

CH3OPOOH3COH3C

CAS RN 32806-04-5

IUPAC Name: diethyl (methoxymethyl)phosphonate
Synonyms: diethyl methoxymethylphosphonate
Molecular Weight (g/mol): 182.16
Molecular Formula: C6H15O4P
InChi Key: IWEOGHGFCXMKFM-UHFFFAOYSA-N
SMILES: CCOP(=O)(COC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results