missing translation for 'onlineSavingsMsg'
Learn More

Diethyl L-(-)-Malate 97.0+%, TCI America™

Catalog No. p-7132679
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13485G 5 g
M134825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13485G Supplier TCI America Supplier No. M13485G

Chemical Identifiers

CAS 691-84-9
Molecular Formula C8H14O5
Molecular Weight (g/mol) 190.20
MDL Number MFCD00210119
InChI Key VKNUORWMCINMRB-LURJTMIESA-N
Synonym L-(-)-Apple Acid Diethyl Ester, L-(-)-Malic Acid Diethyl Ester
PubChem CID 1715085
IUPAC Name 1,4-diethyl (2S)-2-hydroxybutanedioate
SMILES CCOC(=O)C[C@H](O)C(=O)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl L-(-)-Malate