accessibility menu, dialog, popup

UserName

H3COO(S)OHOOCH3

CAS RN 691-84-9

IUPAC Name: 1,4-diethyl (2S)-2-hydroxybutanedioate
Synonyms: 1,4-diethyl (2S)-2-hydroxybutanedioate
Molecular Weight (g/mol): 190.2
Molecular Formula: C8H14O5
InChi Key: VKNUORWMCINMRB-LURJTMIESA-N
SMILES: CCOC(=O)C[C@H](O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results