missing translation for 'onlineSavingsMsg'
Learn More

Diethyl L-Aspartate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128441
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47565G 5 g
D475625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47565G Supplier TCI America Supplier No. D47565G

Chemical Identifiers

CAS 16115-68-7
Molecular Formula C8H16ClNO4
Molecular Weight (g/mol) 225.669
MDL Number MFCD00038910
InChI Key AJOXZAAREAYBQR-RGMNGODLSA-N
Synonym l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
PubChem CID 12416466
IUPAC Name diethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CCOC(=O)CC(C(=O)OCC)N.Cl

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 225.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl L-Aspartate Hydrochloride