accessibility menu, dialog, popup

UserName

CAS RN 16115-68-7

IUPAC Name: 1,4-diethyl 2-aminobutanedioate hydrochloride
Synonyms: 1,4-diethyl 2-aminobutanedioate hydrochloride
Molecular Weight (g/mol): 225.67
Molecular Formula: C8H16ClNO4
InChi Key: AJOXZAAREAYBQR-UHFFFAOYNA-N
SMILES: Cl.CCOC(=O)CC(N)C(=O)OCC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)