Learn More
Diethyl ketomalonate, 95%
CAS: 609-09-6 | C7H10O5 | 174.152 g/mol
Supplier: Thermo Scientific Chemicals A1277114
Description
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.
Solubility
Fully miscible with water.
Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Specifications
| Diethyl ketomalonate | |
| ∼-70°C | |
| 208°C to 210°C | |
| C7H10O5 | |
| MFCD00009121 | |
| 971873 | |
| 14,5913 | |
| Fully miscible with water. | |
| CCOC(=O)C(=O)C(=O)OCC | |
| 174.152 | |
| 174.15 |
| 609-09-6 | |
| 1.142 | |
| >110°C (230°F) | |
| 1.415 | |
| 25 g | |
| Moisture sensitive | |
| diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
| DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
| diethyl 2-oxopropanedioate | |
| 69105 | |
| 95% |
Chemical Identifiers
| 609-09-6 | |
| 174.152 | |
| DBKKFIIYQGGHJO-UHFFFAOYSA-N | |
| 69105 | |
| CCOC(=O)C(=O)C(=O)OCC |
| C7H10O5 | |
| MFCD00009121 | |
| diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester | |
| diethyl 2-oxopropanedioate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-176-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only