missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ketomalonate, 95%

Catalog No. AAA1277106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1277106 5 g
AAA1277114 25 g
AAA1277122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1277106 Supplier Thermo Scientific Chemicals Supplier No. A1277106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 609-09-6 | C7H10O5 | 174.152 g/mol

Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

Solubility
Fully miscible with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.

Chemical Identifiers

CAS 609-09-6
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.152
MDL Number MFCD00009121
InChI Key DBKKFIIYQGGHJO-UHFFFAOYSA-N
Synonym diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester
PubChem CID 69105
IUPAC Name diethyl 2-oxopropanedioate
SMILES CCOC(=O)C(=O)C(=O)OCC

Specifications

Melting Point ∼-70°C
Density 1.142
Boiling Point 208°C to 210°C
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 5 g
Beilstein 971873
Sensitivity Moisture sensitive
Merck Index 14,5913
Solubility Information Fully miscible with water.
Formula Weight 174.15
Percent Purity 95%
Chemical Name or Material Diethyl ketomalonate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-176-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only